D-(+)-Sucrose 99%

Supplier: Thermo Scientific Chemicals

A15583-0C A15583-36
CAAAAA15583-0CEA 434.5 CAD
CAAAAA15583-0C CAAAAA15583-36
D-(+)-Sucrose 99%
D(+)-Sucrose

Used for protein purification in sucrose density gradient studies


Purity: 99% CAS No.: 57-50-1 Molecular Formula: C12H22O11 Formula Weight: 342.30 Odor: Odorless Melting Point: 186-190° Beilstein Reference: 90825 Merck Reference: 14,8881 Optical Rotation: +66.5° (c=10 in water) MDL No.: MFCD00006626

Formula: C₁₂H₂₂O₁₁
Melting Pt: 186…190 °C
Density: 1.580
Storage Temperature: Ambient
MDL Number: MFCD00006626
CAS Number: 57-50-1
EINECS: 200-334-9
Merck Index: 14,8881
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=971b1a9d1f02c06efd63ac665dfb191799fbd5ac&vid=2ddc1affd401bacf02244ae0cb620ff18e440965&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR