Lanthanum(III) chloride heptahydrate 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A15575-0E
A15575-22
A15575-36
CAAAAA15575-0EEA
1845.44
CAD
CAAAAA15575-0E
CAAAAA15575-22
CAAAAA15575-36
Lanthanum(III) chloride heptahydrate 99%
Lanthanum (III) chloride heptahydrate
Purity: 99%
CAS No.: 10025-84-0
Molecular Formula: LaCl3.7H2O
Formula Weight: 371.37 (245.27anhy)
Odor: Odorless
Melting Point: 91° dec.
MDL No.: MFCD00149756
Formula:
LaCl₃·7H₂O MW: 371.37 g/mol Boiling Pt: ? 860 °C (1013 hPa) Melting Pt: 91 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00149756 CAS Number: 10025-84-0 EINECS: 233-237-5 Merck Index: 14,5363 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...