Iron(II) sulfide, tech.

Supplier: Thermo Scientific Chemicals

A15569-0B A15569-0I A15569-30
CAAAAA15569-0BEA 153.1 CAD
CAAAAA15569-0B CAAAAA15569-0I CAAAAA15569-30
Iron(II) sulfide, tech.
Iron (II) sulphide

CAS No.: 1317-37-9 Molecular Formula: FeS Formula Weight: 87.91 Odor: Odorless Melting Point: 1195° Merck Reference: 14,4058 MDL No.: MFCD00011013

Formula: FeS
Melting Pt: 1195 °C
Density: 4.74
Storage Temperature: Ambient
MDL Number: MFCD00011013
CAS Number: 1317-37-9
EINECS: 215-268-6
ADR: 9,III
Merck Index: 14,4058

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR