Nickel(II) nitrate hexahydrate 98%

Supplier: Thermo Scientific Chemicals
Danger

A15540-0B A15540-0I A15540-22 A15540-36 A15540-A4
CAAAAA15540-0BEA 167.48 CAD
CAAAAA15540-0B CAAAAA15540-0I CAAAAA15540-22 CAAAAA15540-36 CAAAAA15540-A4
Nickel(II) nitrate hexahydrate 98%
Nickel (II) nitrate hexahydrate

Purity: 98% CAS No.: 13478-00-7 Molecular Formula: N2NiO6.6H2O Formula Weight: 290.80 (182.70anhy) Odor: Odorless Melting Point: 56.7° Boiling Point: 137° Merck Reference: 14,6513 UN No.: 2725 MDL No.: MFCD00149805

Formula: Ni(NO₃)₂·6H₂O
MW: 290.79 g/mol
Boiling Pt: 137 °C
Melting Pt: 56.7 °C
Density: 2.05
Storage Temperature: Ambient
MDL Number: MFCD00149805
CAS Number: 13478-00-7
EINECS: 236-068-5
UN: 2725
ADR: 5.1 III
Merck Index: 14,6513

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR