Methyl phenylacetate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Phenylacetic acid methyll ester

A15537-0B A15537-0I A15537-30
CAAAA15537-0BEA 121.19 CAD
CAAAA15537-0B CAAAA15537-0I CAAAA15537-30
Methyl phenylacetate 99%
Methyl phenylacetate
Formula: C₉H₁₀O₂
Boiling Pt: 218…220 °C
Melting Pt: 49…50 °C
Density: 1.067
Flash Pt: 85 °C(185 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008453
CAS Number: 101-41-7
EINECS: 202-940-9
Merck Index: 14,7268

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR