Lithium hydroxide monohydrate 98%

Supplier: Thermo Scientific Chemicals
Danger

A15519-0E A15519-22 A15519-36
CAAAAA15519-0EEA 1485.2 CAD
CAAAAA15519-0E CAAAAA15519-22 CAAAAA15519-36
Lithium hydroxide monohydrate 98%
Lithium hydroxide monohydrate

Purity: 98% CAS No.: 1310-66-3 Molecular Formula: LiOH.H2O Formula Weight: 41.96 (23.95anhy) Merck Reference: 14,5534 UN No.: 2680 MDL No.: MFCD00149772

Formula: LiOH·1H₂O
Boiling Pt: 920 °C (1013 hPa)
Melting Pt: 462 °C
Density: 1.51
Storage Temperature: Ambient
MDL Number: MFCD00149772
CAS Number: 1310-66-3
EINECS: 215-183-4
UN: 2680
ADR: 8,II
Merck Index: 14,5534

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR