Indium phosphide 99% (metals basis), polycrystalline lumps 2-8 mm (0.08-0.3 in)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Indium(III) phosphide, Indium phosphide

A15509-03 A15509-06
CAAAAA15509-03EA 151.05 CAD
CAAAAA15509-03 CAAAAA15509-06 CAAAAA15509-14
Indium phosphide 99% (metals basis), polycrystalline lumps 2-8 mm (0.08-0.3 in)
Indium phosphide

Purity: 99.99% CAS No.: 22398-80-7 Molecular Formula: InP Formula Weight: 145.79 Melting Point: 1070° Merck Reference: 14,4953 MDL No.: MFCD00016153

Formula: InP
MW: 145.79 g/mol
Melting Pt: 1062
Density: 4.81
Storage Temperature: Ambient
MDL Number: MFCD00016153
CAS Number: 22398-80-7
EINECS: 244-959-5
Merck Index: 14,4953

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR