Tripropyl orthoformate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,1',1''-[Methylidynetris(oxy)]trispropane, Tri-n-propyl orthoformate, Tripropylorthoformate

A15503-06 A15503-14 A15503-22
CAAAA15503-06EA 53.25 CAD
CAAAA15503-06 CAAAA15503-14 CAAAA15503-22
Tripropyl orthoformate 97%
Tripropyl orthoformate

Purity: 97% CAS No.: 621-76-1 Molecular Formula: HC(OCH2CH2CH3)3 Formula Weight: 190.28 Flash Point: 72°(161°F) Refractive Index: 1.4070 Boiling Point: 106-108°/40mm Beilstein Reference: 1744249 MDL No.: MFCD00015214

Formula: C₁₀H₂₂O₃
MW: 190.28 g/mol
Boiling Pt: 106…108 °C (40 mmHg)
Density: 0.886
Flash Pt: 72 °C (161 °F)
Storage Temperature: Ambient
MDL Number: MFCD00015214
CAS Number: 621-76-1
EINECS: 210-704-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR