Ethyl acrylate 99% stabilized
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Acrylic acid-ethyl ester
Stabilization:
Stabilised with hydroquinone monomethyl ether (4-methoxyphenol, MEHQ) ∼20 ppm
A15493-AE
A15493-AP
CAAAAA15493-AEEA
55.14
CAD
CAAAAA15493-AE
CAAAAA15493-AP
Ethyl acrylate 99% stabilized
Ethyl acrylate
Purity: 99%
CAS No.: 140-88-5
Molecular Formula: C5H8O2
Formula Weight: 100.12
Storage Temperature: 2°C to 8°C
Flash Point: -1°(30°F)
Refractive Index: 1.4060
Odor: Pungent
Melting Point: -71°
Boiling Point: 99-100°
Beilstein Reference: 773866
Merck Reference: 14,3759
UN No.: 1917
MDL No.: MFCD00009188
Formula:
C₅H₈O₂ MW: 100.12 g/mol Boiling Pt: 99…100 °C Melting Pt: –71 °C Density: 0.924 Flash Pt: -1 °C(30 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009188 CAS Number: 140-88-5 EINECS: 205-438-8 UN: 1917 ADR: "3",II Merck Index: 14,3759 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...