4-Fluorobenzylamine 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(4-Fluorophenyl)methanamine
A15486-09
A15486-18
A15486-30
CAAAAA15486-09EA
57.99
CAD
CAAAAA15486-09
CAAAAA15486-18
CAAAAA15486-30
4-Fluorobenzylamine 98+%
4-Fluorobenzylamine
Purity: 98+%
CAS No.: 140-75-0
Molecular Formula: C7H8FN
Formula Weight: 125.15
Flash Point: 66°(150°F)
Refractive Index: 1.5130
Odor: Amine-like
Boiling Point: 182-183°
Beilstein Reference: 636501
UN No.: 2735
MDL No.: MFCD00008120
Formula:
C₇H₈FN MW: 125.15 g/mol Boiling Pt: 182…183 °C Density: 1.095 Flash Pt: 66 °C (150 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008120 CAS Number: 140-75-0 EINECS: 205-430-4 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...