Sodium iodide, (max. 1% H₂O) 99+% (dry weight)
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15480-0E
A15480-22
A15480-36
CAAAAA15480-0EEA
1208.7
CAD
CAAAAA15480-0E
CAAAAA15480-22
CAAAAA15480-36
Sodium iodide, (max. 1% H₂O) 99+% (dry weight)
Sodium iodide
Purity: 99+%
CAS No.: 7681-82-5
Molecular Formula: NaI
Formula Weight: 149.89
Refractive Index: 1.7745
Odor: Odorless
Boiling Point: 1300-1304°
Merck Reference: 14,8631
MDL No.: MFCD00003532
Formula:
NaI Boiling Pt: 1300…1304 °C Melting Pt: 651 °C Density: 3.67 Storage Temperature: Ambient |
MDL Number:
MFCD00003532 CAS Number: 7681-82-5 EINECS: 231-679-3 UN: 3077 ADR: 9,III Merck Index: 14,8631 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...