Hexaamminecobalt trichloride 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Hexaamminecobalt(iii)chloride, Hexammine cobalt(III) chloride, Hexaamminecobalt (III) chloride

A15470-14 A15470-22 A15470-36
CAAAAA15470-14EA 183.28 CAD
CAAAAA15470-14 CAAAAA15470-22 CAAAAA15470-36
Hexaamminecobalt trichloride 99%
Hexaamminecobalt trichloride

Purity: 99% CAS No.: 10534-89-1 Molecular Formula: H18Cl3CoN6 Formula Weight: 267.48 Odor: Odorless Merck Reference: 14,4675 MDL No.: MFCD00036304

Formula: H₁₈CoCl₃N₆
MW: 267.47 g/mol
Density: 1.71
Storage Temperature: Ambient
MDL Number: MFCD00036304
CAS Number: 10534-89-1
EINECS: 234-103-9
Merck Index: 14,4675

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR