1-Bromo-2-methoxyethane 96% stabilised
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Bromoethyl methyl ether
Stabilization:
Stabilised with sodium carbonate (Na₂CO₃)
A15370-06
A15370-14
A15370-22
A15370-36
CAAAAA15370-06EA
41.71
CAD
CAAAAA15370-06
CAAAAA15370-14
CAAAAA15370-22
CAAAAA15370-36
1-Bromo-2-methoxyethane 96% stabilised
1-Bromo-2-methoxyethane
Purity: 96%
CAS No.: 6482-24-2
Molecular Formula: BrCH2CH2OCH3
Formula Weight: 139.00
Storage Temperature: 2°C to 8°C
Flash Point: 28°(82°F)
Refractive Index: 1.4480
Odor: Recognizable
Boiling Point: 109-110°
UN No.: 3271
MDL No.: MFCD00000236
Formula:
BrH₂CCH₂OCH₃ Boiling Pt: 109…110 °C Density: 1.478 Flash Pt: 28 °C (82 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00000236 CAS Number: 6482-24-2 EINECS: 229-339-4 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...