m-Phenylenediamine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: MPD, 1,3-Diaminobenzene, 1,3-Phenylenediamine

A15339-0B A15339-30
CAAAAA15339-0BEA 172.22 CAD
CAAAAA15339-0B CAAAAA15339-30
m-Phenylenediamine 98%
m-Phenylenediamine

MDL: MFCD00007799 Beilstein: 471357 Soluble in water, methanol, acetone, DMF, MEK, dioxane


Purity: 98% CAS No.: 108-45-2 Molecular Formula: C6H8N2 Formula Weight: 108.14 Flash Point: 187°(368°F) Melting Point: 62-66° Boiling Point: 284-287° Solubility: Soluble in water, methanol, acetone, DMF, MEK, dioxane Beilstein Reference: 471357 Merck Reference: 14,7283 UN No.: 1673 MDL No.: MFCD00007799

Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 284…287 °C
Melting Pt: 62…66 °C
Density: 1.139
Flash Pt: 187 °C (368 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007799
CAS Number: 108-45-2
EINECS: 203-584-7
UN: 1673
ADR: 6.1 III
Merck Index: 14,7283

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR