1,4-Diiodobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

A15335-14 A15335-22
CAAAAA15335-14EA 106.33 CAD
CAAAAA15335-14 CAAAAA15335-22
1,4-Diiodobenzene 98%
1,4-Diiodobenzene

Purity: 98% CAS No.: 624-38-4 Molecular Formula: C6H4I2 Formula Weight: 329.91 Melting Point: 128-132° Boiling Point: 285° MDL No.: MFCD00001054

Formula: C₆H₄I₂
MW: 329.91 g/mol
Boiling Pt: 285 °C
Melting Pt: 128…132 °C
Density: 2,88 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001054
CAS Number: 624-38-4
EINECS: 210-842-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR