2-Pyrrolidone 99%

Supplier: Thermo Scientific Chemicals
Warning

A15332-36 A15332-A3
CAAAA15332-36EA 63.99 CAD
CAAAA15332-36 CAAAA15332-A3
2-Pyrrolidone 99%
2-Pyrrolidone

Purity: 99% CAS No.: 616-45-5 Molecular Formula: C4H7NO Formula Weight: 85.11 Flash Point: 110°(230°F) Refractive Index: 1.4875 Melting Point: 23-25° Boiling Point: 245° Merck Reference: 14,8016 MDL No.: MFCD00005270

Formula: C₄H₇NO
Boiling Pt: 245 °C
Melting Pt: 23…25 °C
Density: 1.12
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005270
CAS Number: 616-45-5
EINECS: 210-483-1
Merck Index: 14,8016

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR