4-Methoxyphenyl isothiocyanate 98%

Supplier: Thermo Scientific Chemicals
Danger

A15331-04 A15331-09 A15331-18
CAAAAA15331-04EA 44.87 CAD
CAAAAA15331-04 CAAAAA15331-09 CAAAAA15331-18
4-Methoxyphenyl isothiocyanate 98%
4-Methoxyphenyl isothiocyanate

Purity: 98% CAS No.: 2284-20-0 Molecular Formula: C8H7NOS Formula Weight: 165.22 Flash Point: 110°(230°F) Refractive Index: 1.6489 Odor: Pungent Melting Point: 18-20° Boiling Point: 279-281° Beilstein Reference: 606967 UN No.: 2922 MDL No.: MFCD00011676

Formula: C₈H₇NOS
Boiling Pt: 279…281 °C
Melting Pt: 18…20 °C
Density: 1.196
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011676
CAS Number: 2284-20-0
EINECS: 218-921-3
UN: 1760
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR