3,5-Dichlorophenol 98%

Supplier: Thermo Scientific Chemicals
Warning

A15328-06 A15328-14 A15328-18
CAAAAA15328-06EA 99.7 CAD
CAAAAA15328-06 CAAAAA15328-14 CAAAAA15328-18
3,5-Dichlorophenol 98%
3,5-Dichlorophenol

Purity: 98% CAS No.: 591-35-5 Molecular Formula: C6H4Cl2O Formula Weight: 163.00 Odor: Phenol-like Melting Point: 63-68° Boiling Point: 233° Beilstein Reference: 1906458 UN No.: 2020 MDL No.: MFCD00002259

Formula: C₆H₄Cl₂O
Boiling Pt: 233 °C
Melting Pt: 63…68 °C
Storage Temperature: Ambient
MDL Number: MFCD00002259
CAS Number: 591-35-5
EINECS: 209-714-9
UN: 2020
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR