Methyl-2-bromo-5-methoxybenzoate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15322-06
A15322-14
CAAAA15322-06EA
106.33
CAD
CAAAA15322-06
CAAAA15322-14
Methyl-2-bromo-5-methoxybenzoate 98%
Purity: 98%
CAS No.: 35450-36-3
Molecular Formula: C9H9BrO3
Formula Weight: 245.08
Flash Point:
Formula:
C₉H₉BrO₃ Boiling Pt: 271 °C Density: 1.52 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00051594 CAS Number: 35450-36-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...