4-Methylbenzyl alcohol 99%

Supplier: Thermo Scientific Chemicals
Warning

A15315-14 A15315-22 A15315-36
CAAAAA15315-14EA 85.48 CAD
CAAAAA15315-14 CAAAAA15315-22 CAAAAA15315-36
4-Methylbenzyl alcohol 99%
4-Methylbenzyl alcohol

Purity: 99% CAS No.: 589-18-4 Molecular Formula: C8H10O Formula Weight: 122.17 Melting Point: 57-61° Boiling Point: 217° MDL No.: MFCD00004664

Formula: C₈H₁₀O
Boiling Pt: 217 °C
Melting Pt: 57…61 °C
Density: 0,978
Storage Temperature: Ambient
MDL Number: MFCD00004664
CAS Number: 589-18-4
EINECS: 209-639-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR