Potassium peroxodisulfate 97%

Supplier: Thermo Scientific Chemicals
Danger

A15310-0E A15310-36
CAAAAA15310-0EEA 387.1 CAD
CAAAAA15310-0E CAAAAA15310-36
Potassium peroxodisulfate 97%
Potassium peroxodisulphate

Purity: 97% CAS No.: 7727-21-1 Molecular Formula: K2O8S2 Formula Weight: 270.33 Odor: Odorless Melting Point: 100° dec. UN No.: 1492 MDL No.: MFCD00011386

Formula: K₂S₂O₈
Melting Pt: 100 °C (dec.)
Density: 2.477
Storage Temperature: Ambient
MDL Number: MFCD00011386
CAS Number: 7727-21-1
EINECS: 231-781-8
UN: 1492
ADR: 5.1,III
Merck Index: 14,7656

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR