Ammonium dihydrogen phosphate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Ammonium phosphate, monobasic, Ammonium Phosphate Monobasic, Ammonium dihydrogenphosphate

A15283-0C A15283-0E A15283-36
CAAAAA15283-0CEA 620.94 CAD
CAAAAA15283-0C CAAAAA15283-0E CAAAAA15283-36
Ammonium dihydrogen phosphate 98%
Ammonium dihydrogen phosphate

Purity: 98% CAS No.: 7722-76-1 Molecular Formula: NH4H2PO4 Formula Weight: 115.03 Odor: Odorless Melting Point: 198-204° MDL No.: MFCD00003396

Formula: NH₄H₂PO₄
MW: 115.03 g/mol
Melting Pt: 198…204 °C
Density: 1.803
Storage Temperature: Ambient
MDL Number: MFCD00003396
CAS Number: 7722-76-1
EINECS: 231-764-5
Merck Index: 14,543

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR