Boron trifluoride diethyl ether complex 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A15275-AE
A15275-AP
A15275-AU
CAAAAA15275-AEEA
68.98
CAD
CAAAAA15275-AE
CAAAAA15275-AP
CAAAAA15275-AU
Boron trifluoride diethyl ether complex 98+%
Boron trifluoride diethyl ether complex
Purity: 98+%
CAS No.: 109-63-7
Molecular Formula: C4H10BF3O
Formula Weight: 141.93
Flash Point: 47°(116°F)
Refractive Index: 1.3440
Odor: Acrid
Melting Point: -60°
Boiling Point: 125-126°
Beilstein Reference: 3909607
Merck Reference: 14,1350
UN No.: 2604
MDL No.: MFCD00013194
Formula:
C₄H₁₀BF₃O Boiling Pt: 125…126 °C Melting Pt: –60 °C Density: 1.130 Flash Pt: 47 °C(116 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00013194 CAS Number: 109-63-7 EINECS: 203-689-8 UN: 2604 ADR: "8",I Merck Index: 14,1350 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...