3-Bromobenzoyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: m-Bromobenzoyl chloride, 3-Bromobenzoylchloride

A15256-30 A15256-09 A15256-18
CAAAAA15256-30EA 917.98 CAD
CAAAAA15256-30 CAAAA15256-09 CAAAA15256-18
3-Bromobenzoyl chloride 98%
3-Bromobenzoyl chloride

Purity: 98% CAS No.: 1711-09-7 Molecular Formula: C7H4BrClO Formula Weight: 219.47 Flash Point: 107°(224°F) Refractive Index: 1.5950 Boiling Point: 247-251° Beilstein Reference: 1635505 UN No.: 3265 MDL No.: MFCD00000669

Formula: C₇H₄BrClO
Boiling Pt: 247…251 °C
Density: 1.666
Flash Pt: 107 °C (224 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000669
CAS Number: 1711-09-7
EINECS: 216-978-9
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR