Triisopropyl phosphite 90+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A15247-AK
A15247-AP
CAAAAA15247-AKEA
162.74
CAD
CAAAAA15247-AK
CAAAAA15247-AP
Triisopropyl phosphite 90+%
Triisopropyl phosphite
Purity: 90+%
CAS No.: 116-17-6
Molecular Formula: [(CH3)2CHO]3P
Formula Weight: 208.24
Flash Point: 46°(114°F)
Refractive Index: 1.4110
Odor: Characteristic
Boiling Point: 63-64°/11mm
Beilstein Reference: 1701528
UN No.: 1993
MDL No.: MFCD00008874
Formula:
C₉H₂₁O₃P Boiling Pt: 63…64 °C/11 mm Density: 0.908 Flash Pt: 46 °C (114 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008874 CAS Number: 116-17-6 EINECS: 204-130-0 UN: 3278 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...