Octamethylcyclotetrasiloxane 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15240-22
A15240-36
CAAAAA15240-22EA
148.05
CAD
CAAAAA15240-22
CAAAAA15240-36
Octamethylcyclotetrasiloxane 98%
Octamethylcyclotetrasiloxane
Purity: 98%
CAS No.: 556-67-2
Molecular Formula: C8H24O4Si4
Formula Weight: 296.62
Flash Point: 56°(132°F)
Refractive Index: 1.3960
Melting Point: 17-18°
Boiling Point: 175-176°
Beilstein Reference: 1787074
Merck Reference: 14,6747
UN No.: 1993
MDL No.: MFCD00003269
Formula:
[-Si(CH₃)₂O-]₄ Boiling Pt: 175…176 °C Melting Pt: 17…18 °C Density: 0.956 Flash Pt: 56 °C (132 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003269 CAS Number: 556-67-2 EINECS: 209-136-7 UN: 1993 ADR: 3,III Merck Index: 14,6747 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...