Tetraethylammonium chloride hydrate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tetrylammonium chloride hydrate

A15215-14 A15215-22 A15215-36
CAAAAA15215-14EA 53.72 CAD
CAAAAA15215-14 CAAAAA15215-22 CAAAAA15215-36
Tetraethylammonium chloride hydrate 98%
Tetraethylammonium chloride

Tetraethylammonium chloride monohydrate is used as pharmaceutical intermediate.


Purity: 98% CAS No.: 68696-18-4 Molecular Formula: (CH3CH2)4NCl.H2O Formula Weight: 183.72 (165.71anhy) Melting Point: ca 110° Beilstein Reference: 3563247 Merck Reference: 14,9200 MDL No.: MFCD00149992


Caution: Stable under recommended storage conditions. Incompatible with oxidizing agents.

Formula: C₈H₂₀ClN·xH₂O
MW: 165.71 g/mol (anhy)
Melting Pt: ∼ 110 °C
Density: 1.0801
Storage Temperature: Ambient
MDL Number: MFCD00149992
CAS Number: 56-34-8
EINECS: 200-267-5
Merck Index: 14,9200

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR