3-Bromo-4-fluorobenzaldehyde 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15198-06
A15198-14
CAAAAA15198-06EA
70.15
CAD
CAAAAA15198-06
CAAAAA15198-14
3-Bromo-4-fluorobenzaldehyde 98%
3-Bromo-4-fluorobenzaldehyde
Purity: 98%
CAS No.: 77771-02-9
Molecular Formula: C7H4BrFO
Formula Weight: 203.01
Flash Point:
Formula:
C₇H₄BrFO Boiling Pt: 138…139 °C/2.5 mm Melting Pt: 29…33 °C Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00042186 CAS Number: 77771-02-9 EINECS: 278-764-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...