6-Methylanthranil acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Amino-6-methylbenzoic acid

A15187-06 A15187-14 A15187-22
CAAAAA15187-06EA 137.3 CAD
CAAAAA15187-06 CAAAAA15187-14 CAAAAA15187-22
6-Methylanthranil acid 98%
6-Methylanthranil acid

Purity: 98% CAS No.: 4389-50-8 Molecular Formula: C8H9NO2 Formula Weight: 151.16 Melting Point: ca 127° dec. Beilstein Reference: 2803391

Formula: C₈H₉NO₂
MW: 151.16 g/mol
Boiling Pt: 315
Melting Pt: ∼ 127 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00007809
CAS Number: 4389-50-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR