Iron(II) sulfate heptahydrate 98%

Supplier: Thermo Scientific Chemicals
Warning

A15178-0C A15178-0E A15178-36
CAAAAA15178-0CEA 586.18 CAD
CAAAAA15178-0C CAAAAA15178-0E CAAAAA15178-36
Iron(II) sulfate heptahydrate 98%
Iron (II) sulphate heptahydrate

Purity: 98% CAS No.: 7782-63-0 Molecular Formula: FeO4S.7H2O Formula Weight: 278.02 (151.91anhy) Melting Point: 64° -6H2O Merck Reference: 14,4057 MDL No.: MFCD00149719

Formula: FeSO₄·7H₂O
Melting Pt: 64 °C (–H₂O of crystall.)
Density: 1.897
Storage Temperature: Ambient
MDL Number: MFCD00149719
CAS Number: 7782-63-0
EINECS: 231-753-5
Merck Index: 14,4057

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR