EDTA disodium salt dihydrate 99+%

Supplier: Thermo Scientific Chemicals
Warning

A15161-0B A15161-0I A15161-30
CAAAAA15161-0BEA 268.54 CAD
CAAAAA15161-0B CAAAAA15161-0I CAAAAA15161-30
EDTA disodium salt dihydrate 99+%
EDTA disodium salt dihydrate

Purity: 99+% CAS No.: 6381-92-6 Molecular Formula: C10H14N2Na2O8.2H2O Formula Weight: 372.24 (336.22anhy) Melting Point: 248° dec. Beilstein Reference: 3900609

Formula: C₁₀H₁₄N₂Na₂O₈·2H₂O
Melting Pt: 248 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00150037
CAS Number: 6381-92-6
EINECS: 205-358-3
Merck Index: 13,03543
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f89002087c5d4d84af5dded62c87287b1774fab&vid=0ef4c3febbcc8a20569606067799db31f2540748&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR