Ammonium hexachlorostannate(IV) 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Diammonium hexachlorostannate

A15152-18 A15152-30
CAAAAA15152-0BEA 610.59 CAD
CAAAAA15152-0B CAAAAA15152-18 CAAAAA15152-30
Ammonium hexachlorostannate(IV) 98%
Ammonium hexachlorostannate(IV)

Purity: 98% CAS No.: 16960-53-5 Molecular Formula: (NH4)2SnCl6 Formula Weight: 367.48 Melting Point: dec. MDL No.: MFCD00036340

Formula: H₈Cl₆N₂Sn
MW: 367.5 g/mol
Melting Pt: Dec.
Density: 2.4
Storage Temperature: Ambient
MDL Number: MFCD00036340
CAS Number: 16960-53-5
EINECS: 241-033-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR