3,5-Dichlorophenylhydrazinium chloride 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-(3,5-Dichlorophenyl)hydrazine hydrochloride, 3,5-Dichlorophenylhydrazine hydrochloride
A15127-14
A15127-06
CAAAAA15127-14EA
461.36
CAD
CAAAAA15127-14
CAAAA15127-03
CAAAA15127-06
3,5-Dichlorophenylhydrazinium chloride 97%
3,5-Dichlorophenylhydrazinium chloride
Purity: 97%
CAS No.: 63352-99-8
Molecular Formula: C6H6Cl2N2.HCl
Formula Weight: 213.50
Melting Point: ca 204° dec.
Beilstein Reference: 4208459
Formula:
C₆H₇Cl₃N₂ MW: 213.49 g/mol Melting Pt: ∼ 204 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00012938 CAS Number: 63352-99-8 EINECS: 264-109-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...