Mesityl oxide ≥90% remainder 4-methyl-4-penten-2-one
Supplier: Thermo Scientific Chemicals
|
Warning
|
A15111-AE
A15111-AP
A15111-AK
A15111-AU
CAAAAA15111-AEEA
86.9
CAD
CAAAAA15111-AE
CAAAAA15111-AP
CAAAA15111-AK
CAAAA15111-AU
Mesityl oxide ≥90% remainder 4-methyl-4-penten-2-one
Mesityl oxide
Purity: 90+%
CAS No.: 141-79-7
Molecular Formula: C6H10O
Formula Weight: 98.15
Flash Point: 30°(86°F)
Physical Form: Clear liquid
Density: 0.856
Refractive Index: 1.4440
Appearance: Colorless
Odor: honigähnlich
Melting Point: -59°
Boiling Point: 129-130°
Beilstein Reference: 1361550
Merck Reference: 14,5908
UN No.: 1229
MDL No.: MFCD00008900
Formula:
C₆H₁₀O Boiling Pt: 129…130 °C Melting Pt: –59 °C Density: 0.856 Flash Pt: 30 °C(86 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008900 CAS Number: 141-79-7 EINECS: 205-502-5 UN: 1229 ADR: "3",III Merck Index: 14,5908 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...