Pivalaldehyde (trimethylacetaldehyde) 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A15013-06
A15013-14
A15013-22
CAAAAA15013-06EA
110.44
CAD
CAAAAA15013-06
CAAAAA15013-14
CAAAAA15013-22
Pivalaldehyde (trimethylacetaldehyde) 95%
Pivalaldehyde (Trimethylacetaldehyde)
Purity: 95%
CAS No.: 630-19-3
Molecular Formula: C5H10O
Formula Weight: 86.13
Storage Temperature: 2°C to 8°C
Flash Point: -15°(5°F)
Refractive Index: 1.3794
Odor: Pleasant
Melting Point: 6°
Boiling Point: 74-76°
Beilstein Reference: 506060
UN No.: 1989
MDL No.: MFCD00006962
Formula:
C₅H₁₀O Boiling Pt: 74…76 °C Melting Pt: 6 °C Density: 0.775 Flash Pt: –15 °C (5 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006962 CAS Number: 630-19-3 EINECS: 211-134-6 UN: 1989 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...