Ammonium chloride 98+%

Supplier: Thermo Scientific Chemicals
Warning

A15000-0B A15000-0I A15000-30
CAAAAA15000-0BEA 76.63 CAD
CAAAAA15000-0B CAAAAA15000-0I CAAAAA15000-30
Ammonium chloride 98+%
Ammonium chloride

Purity: 98+% CAS No.: 12125-02-9 Molecular Formula: H4ClN Formula Weight: 53.49 Refractive Index: 1.642 Odor: Odorless Beilstein Reference: 4371014 Merck Reference: 14,509 MDL No.: MFCD00011420

Formula: NH₄Cl
Melting Pt: 340 °C (subl.)
Density: 1.53
Storage Temperature: Ambient
MDL Number: MFCD00011420
CAS Number: 12125-02-9
EINECS: 235-186-4
Merck Index: 14,509
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e45c4909ae60875d5b9dca2f28f54f5c8028e64&vid=389479ce9f26d046837e12f5c47dd4bc7d47ec2b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR