Calcium propionate hydrate 97%

Supplier: Thermo Scientific Chemicals

A14994-0E A14994-36
CAAAAA14994-0EEA 155.63 CAD
CAAAAA14994-0E CAAAAA14994-36
Calcium propionate hydrate 97%
Calcium propionate hydrate

Purity: 97% CAS No.: 4075-81-4 Molecular Formula: (CH3CH2COO)2Ca.xH2O Formula Weight: 186.22(anhy) Melting Point: 300° Merck Reference: 14,1698 MDL No.: MFCD00167354

Formula: (CH₃CH₂COO)₂Ca·xH₂O
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00167354
CAS Number: 4075-81-4
EINECS: 223-795-8
Merck Index: 14,1698

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR