4-Chlorophenyl isocyanate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A14966-06
A14966-14
A14966-22
CAAAAA14966-06EA
42.98
CAD
CAAAAA14966-06
CAAAAA14966-14
CAAAAA14966-22
4-Chlorophenyl isocyanate 98%
4-Chlorophenyl isocyanate
Purity: 98%
CAS No.: 104-12-1
Molecular Formula: C7H4ClNO
Formula Weight: 153.57
Storage Temperature: 2°C to 8°C
Flash Point: 90°(194°F)
Refractive Index: 1.5618
Odor: Pungent
Melting Point: 29-31°
Boiling Point: 203-204°
Beilstein Reference: 386235
UN No.: 2811
MDL No.: MFCD00002024
Formula:
C₇H₄ClNO Boiling Pt: 203…204 °C Melting Pt: 29…31 °C Density: 1.200 Flash Pt: 90 °C (194 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00002024 CAS Number: 104-12-1 EINECS: 203-176-9 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...