Tetraethyl orthosilicate (TEOS) 98%

Supplier: Thermo Scientific Chemicals
Warning

A14965-0F A14965-AE A14965-AP
CAAAAA14965-0FEA 263.86 CAD
CAAAAA14965-0F CAAAAA14965-AE CAAAAA14965-AP
Tetraethyl orthosilicate (TEOS) 98%
Tetraethyl orthosilicate (TEOS)

Purity: 98% CAS No.: 78-10-4 Molecular Formula: C8H20O4Si Formula Weight: 208.33 Flash Point: 46°(116°F) Physical Form: Clear liquid Density: 0.934 Refractive Index: 1.3830 Sensitivity: Moisture Sensitive Appearance: Colorless Odor: Pleasant Melting Point: -85° Boiling Point: 168° Beilstein Reference: 1422225 Merck Reference: 14,3851 UN No.: 1292 MDL No.: MFCD00009062

Formula: Si(OC₂H₅)₄
Boiling Pt: 168 °C
Melting Pt: –85 °C
Density: 0.934
Flash Pt: 46 °C (116 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009062
CAS Number: 78-10-4
EINECS: 201-083-8
UN: 1292
ADR: 3,III
Merck Index: 14,3851

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR