1-Amino-2,2,2-trifluoroethane hydrochloride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14904-09
A14904-18
A14904-22
CAAAAA14904-09EA
128.45
CAD
CAAAAA14904-09
CAAAAA14904-18
CAAAAA14904-22
1-Amino-2,2,2-trifluoroethane hydrochloride 98%
1-Amino-2,2,2-trifluoroethane hydrochloride
Purity: 98%
CAS No.: 373-88-6
Molecular Formula: C2F3H4N.HCl
Formula Weight: 135.52
Melting Point: 218° subl.
Beilstein Reference: 3652103
Formula:
C₂H₅ClF₃N Melting Pt: 218 °C (subl.) Storage Temperature: Ambient |
MDL Number:
MFCD00012875 CAS Number: 373-88-6 EINECS: 206-771-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...