(Chloromethyl)dimethylchlorosilane 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
CMDMCS, Chloro(chloromethyl)dimethylsilane
A14899-09
A14899-14
A14899-22
CAAAAA14899-09EA
40.29
CAD
CAAAAA14899-09
CAAAA14899-14
CAAAA14899-22
(Chloromethyl)dimethylchlorosilane 98%
(Chloromethyl)dimethylchlorosilane
Purity: 98%
CAS No.: 1719-57-9
Molecular Formula: C3H8Cl2Si
Formula Weight: 143.09
Flash Point: 21°(69°F)
Refractive Index: 1.4370
Odor: Acrid
Boiling Point: 110-113°
Beilstein Reference: 1731933
UN No.: 2985
MDL No.: MFCD00000875
Formula:
C₃H₈Cl₂Si MW: 143.09 g/mol Boiling Pt: 110…113 °C Density: 1.082 Flash Pt: 21 °C(69 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000875 CAS Number: 1719-57-9 EINECS: 217-006-6 UN: 2985 ADR: "3",II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...