3,5-Lutidine 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A14877-AC
A14877-AE
A14877-AP
CAAAAA14877-ACEA
42.66
CAD
CAAAAA14877-AC
CAAAAA14877-AE
CAAAAA14877-AP
3,5-Lutidine 99%
3,5-Lutidine
MDL: MFCD00006404
Beilstein: 105682
Sol
Purity: 99%
CAS No.: 591-22-0
Molecular Formula: C7H9N
Formula Weight: 107.16
Flash Point: 53°(127°F)
Refractive Index: 1.5050
Odor: Unpleasant
Melting Point: -9°
Boiling Point: 169-170°
Solubility: Sol
Beilstein Reference: 105682
UN No.: 1992
MDL No.: MFCD00006404
Formula:
C₇H₉N Boiling Pt: 169…170 °C Melting Pt: –9 °C Density: 0.941 Flash Pt: 53 °C (127 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006404 CAS Number: 591-22-0 EINECS: 209-708-6 UN: 2920 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...