4-Benzyloxyphenol 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Monobenzone4-Hydroxyphenyl benzyl etherHydroquinone monobenzyl ether4-(Benzyloxy)phenol

A14843-0B A14843-14 A14843-22
CAAAAA14843-0BEA 1303.5 CAD
CAAAAA14843-0B CAAAAA14843-14 CAAAAA14843-22
4-Benzyloxyphenol 98+%
4-Benzyloxyphenol

Purity: 98+% CAS No.: 103-16-2 Molecular Formula: C13H12O2 Formula Weight: 200.24 Melting Point: 118-122° Beilstein Reference: 1958305 Merck Reference: 14,6248 MDL No.: MFCD00002333

Formula: C₁₃H₁₂O₂
Boiling Pt: 328 °C (1013 hPa)
Melting Pt: 118…122 °C
Density: 1.260
Storage Temperature: Ambient
MDL Number: MFCD00002333
CAS Number: 103-16-2
EINECS: 203-083-3
Merck Index: 14,6248

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR