2-Acetyl-5-bromothiophene 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(5-Bromo-2-thienyl)-1-ethanone

A14826-09 A14826-18
CAAAAA14826-09EA 65.25 CAD
CAAAAA14826-09 CAAAAA14826-18 CAAAAA14826-30
2-Acetyl-5-bromothiophene 99%
2-Acetyl-5-bromothiophene

Purity: 99% CAS No.: 5370-25-2 Molecular Formula: C6H5BrOS Formula Weight: 205.07 Melting Point: 93-96° Boiling Point: 103°/4mm MDL No.: MFCD00014528

Formula: C₆H₅BrOS
MW: 205.08 g/mol
Boiling Pt: 253
Melting Pt: 93…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00014528
CAS Number: 5370-25-2
EINECS: 226-363-7
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR