3-Chlorophenylhydrazinium chloride 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14770-30
A14770-09
A14770-18
CAAAAA14770-30EA
887.96
CAD
CAAAAA14770-30
CAAAA14770-09
CAAAA14770-18
3-Chlorophenylhydrazinium chloride 97%
3-Chlorophenylhydrazinium chloride
Purity: 97%
CAS No.: 2312-23-4
Molecular Formula: C6H7ClN2.HCl
Formula Weight: 179.05
Odor: Odorless
Melting Point: ca 242° dec.
Beilstein Reference: 3565828
MDL No.: MFCD00012935
Formula:
C₆H₈Cl₂N₂ Melting Pt: ∼ 242 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD02656655 CAS Number: 2312-23-4 EINECS: 219-005-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...