Diethylene glycol 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,2'-Oxybis(ethan-1-ol), Digol, 2,2'-Oxybisethanol

A14728-0C A14728-0E A14728-36
CAAAAA14728-0CEA 334.96 CAD
CAAAAA14728-0C CAAAAA14728-0E CAAAAA14728-36
Diethylene glycol 99%
Diethylene glycol

Purity: 99% CAS No.: 111-46-6 Molecular Formula: C4H10O3 Formula Weight: 106.12 Flash Point: 143°(289°F) Physical Form: Clear liquid Density: 1.118 Refractive Index: 1.4470 Sensitivity: Hygroscopic Appearance: Colorless Odor: Nearly odorless Melting Point: -10° Boiling Point: 245-246° Beilstein Reference: 969209 Merck Reference: 14,3119 MDL No.: MFCD00002882

Formula: C₄H₁₀O₃
MW: 106.12 g/mol
Boiling Pt: 245
Melting Pt: -10
Density: 1.118
Flash Pt: 143 °C (289 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002882
CAS Number: 111-46-6
EINECS: 203-872-2
Merck Index: 14,3119
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f023bd53ecff654f6d6d398b3ee2a1bac87d56f&vid=25d8943f9c247b4203b1e1bcba59bf77dfdd9158&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR