Pentadecanoic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14664-09
A14664-18
A14664-30
CAAAAA14664-09EA
112.5
CAD
CAAAAA14664-09
CAAAAA14664-18
CAAAAA14664-30
Pentadecanoic acid 99%
Pentadecanoic acid
Purity: 99%
CAS No.: 1002-84-2
Molecular Formula: C15H30O2
Formula Weight: 242.40
Flash Point: 110°(230°F)
Melting Point: 51-54°
Boiling Point: 257°/100mm
Beilstein Reference: 1773831
MDL No.: MFCD00002745
Formula:
C₁₅H₃₀O₂ Boiling Pt: 257 °C/100 mm Melting Pt: 51…54 °C Density: 0,8496 g/cm³ (70 °C) Flash Pt: 110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00002745 CAS Number: 1002-84-2 EINECS: 213-693-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...