Tin(II) chloride dihydrate 98%

Supplier: Thermo Scientific Chemicals
Danger

A14610-0E A14610-22 A14610-36
CAAAAA14610-0EEA 695.2 CAD
CAAAAA14610-0E CAAAAA14610-22 CAAAAA14610-36
Tin(II) chloride dihydrate 98%
Tin (II) chloride dihydrate

Purity: 98% CAS No.: 10025-69-1 Molecular Formula: Cl2Sn.2H2O Formula Weight: 225.65 (189.62anhy) Melting Point: 42-46° Merck Reference: 14,8783 MDL No.: MFCD00149863

Formula: SnCl₂·2H₂O
Boiling Pt: 623 °C (1013 hPa)
Melting Pt: 42…46 °C
Density: 2.71
Storage Temperature: Ambient
MDL Number: MFCD00149863
CAS Number: 10025-69-1
EINECS: 600-045-1
UN: 3260
ADR: 8,III
Merck Index: 14,8783

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR