Maleic acid 98+%

Supplier: Thermo Scientific Chemicals
Danger

A14596-0B A14596-0I A14596-30
CAAAAA14596-0BEA 115.34 CAD
CAAAAA14596-0B CAAAAA14596-0I CAAAAA14596-30
Maleic acid 98+%
Maleic acid

Purity: 98+% CAS No.: 110-16-7 Molecular Formula: C4H4O4 Formula Weight: 116.07 Flash Point: 127°(260°F) Melting Point: 132-136° Beilstein Reference: 605762 Merck Reference: 14,5703 MDL No.: MFCD00063177

Formula: C₄H₄O₄
MW: 116.07 g/mol
Boiling Pt: 256 °C (1013 hPa)
Melting Pt: 132…136 °C
Density: 1.59
Flash Pt: 127 °C (260 °F)
Storage Temperature: Ambient
MDL Number: MFCD00063177
CAS Number: 110-16-7
EINECS: 203-742-5
UN: 3261
ADR: 8,III
Merck Index: 14,5703
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3a1ea1f5bca5d5cf7fd40530f642f47db88908a&vid=1ac7f8cee2cd0433dbfb6e9271d41fe6ecc382c1&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR