DL-α-Tocopherol acetate 97%
Supplier: Thermo Scientific Chemicals
A14505-14
A14505-22
A14505-36
CAAAA14505-14EA
66.41
CAD
CAAAA14505-14
CAAAA14505-22
CAAAA14505-36
DL-α-Tocopherol acetate 97%
DL-α-Tocopherol acetate
Purity: 97%
CAS No.: 7695-91-2
Molecular Formula: C31H52O3
Formula Weight: 472.76
Storage Temperature: 2°C to 8°C
Flash Point: 210°(410°F)
Refractive Index: 1.4960
Melting Point: -28°
Boiling Point:
Formula:
C₃₁H₅₂O₃ Boiling Pt: >300 °C Melting Pt: –28 °C Density: 0.960 Flash Pt: 210 °C(410 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00072042 CAS Number: 7695-91-2 EINECS: 231-710-0 Merck Index: 14,9495 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...